missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIF1 gamma Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA578496
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Positive Control: MCF-7, MDA-MB-231 Predicted Reactivity: Mouse (97%), Xenopus laevis (91%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TIF1 gamma is a member of the Transcriptional Intermediary Factor 1 family, which serve as coactivators and corepressors. Its structure is similar to TIF1 alpha and TIF1 beta, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TIF1 gamma functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells.
Specifica
| TIF1 gamma | |
| Polyclonal | |
| Unconjugated | |
| TRIM33 | |
| 8030451N04Rik; AI413936; E3 ubiquitin-protein ligase TRIM33; Ecto; Ectodermin homolog; Kiaa1113; Protein Rfg7; PTC7; RET-fused gene 7 protein; RFG7; RING-type E3 ubiquitin transferase TRIM33; TF1G; TIF 1-g; Tif1g; TIF1GAMMA; TIF1-gamma; TIFGAMMA; Transcription intermediary factor 1-gamma; transcriptional intermediary factor 1 gamma; Trim33; tripartite motif containing 33; tripartite motif protein 33; tripartite motif-containing 33; tripartite motif-containing protein 33 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51592 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.21 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9UPN9 | |
| TRIM33 | |
| Recombinant protein encompassing a sequence within the center region of Human TIF1 gamma. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto