Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator with Standard Optics
Click to view available options
:
Comprator with Standard Optics
Descrizione
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
- Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
Specifica
Specifica
| Descrizione | Dia.: 13.5mm |
| Da utilizzare con (applicazione) | Water Quality Testing |
| Da utilizzare con (apparecchiatura) | Checkit Comprator Tests |
| Include | Carry case with comparator, vials, discs, reagents and method instructions for all tests |
| Tipo | Checkit Comprator with Standard Optics |
Titolo del prodotto
Ser du en mulighed for forbedring?Del en indholdskorrektion