Learn More
Lovibond™ Tintometer™ Chlorine Dioxide (DPD No. 1 and Glycine) Reagent Pack
Click to view available options
Quantità:
250 Tablets
500 Tablets
Dimensione della confezione:
pezzi 100
pezzi 250
Descrizione
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifica
Specifica
| Da utilizzare con (applicazione) | Photometry Tablet Reagents |
| Da utilizzare con (apparecchiatura) | Test kits, colorimeters and spectrophotometers |
| Quantità | 250 Tablets |
| Tipo | Chlorine Dioxide (DPD No. 1 and Glycine) Reagent Pack |