Learn More
Lovibond™ Tintometer™ Standard Solution Ammonia, 1.3 mg/L NH4 = 1.0mg/L N
Click to view available options
:
Standard Solution Ammonia
Descrizione
- Standard Solution Ammonia
- 1.3 mg/L NH4 = 1.0mg/L N
- For use with Lovibond test kits, colorimeters and spectrophotometers
Specifica
Specifica
| Descrizione | 1.3 mg/L NH4 = 1.0mg/L N |
| Da utilizzare con (apparecchiatura) | Test Kits, Colorimeters and Spectrophotometers |
| Tipo | Amonia Solution |