Learn More
Thermo Scientific Chemicals Tiotropium bromide
Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Click to view available options
Quantità:
250 mg
1 g
Dimensione della confezione:
1g
250mg
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Materiale o nome chimico | Tiotropium bromide |
| Quantità | 1 g |
| Solubilità | Solubility aq. soln.: slightly soluble. |
| Punto di fusione | 218.0°C to 220.0°C |
| Peso formulazione | 472.41 |
| InChI Key | DQHNAVOVODVIMG-CJTHRXGHSA-M |
| SMILES | [Br-].C[N+]1(C)[C@H]2CC(C[C@@H]1[C@@H]1O[C@H]21)OC(=O)C(O)(C1=CC=CS1)C1=CC=CS1 |
| IUPAC Name | (1R,2S,4R,5S)-7-{[2-hydroxy-2,2-bis(thiophen-2-yl)acetyl]oxy}-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium bromide |
| Molecular Weight (g/mol) | 472.41 |