Learn More
Sampling Systems™ Tip for Unit Dose Sleeve Sampler
Codice prodotto. 15889281 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Lunghezza (sistema metrico):
9.5 mm
11.5 mm
13.5 mm
14.5 mm
15.5 mm
17.5 mm
18.5 mm
19.5 mm
21.5 mm
22.5 mm
23.5 mm
25.5 mm
26.5 mm
27.5 mm
29.5 mm
30.5 mm
31.5 mm
34.5 mm
35.5 mm
38 mm
41.5 mm
45.5 mm
49 mm
52.5 mm
54 mm
Capacità (sistema metrico):
0.1 mL
0.2 mL
0.3 mL
0.4 mL
0.5 mL
0.6 mL
0.7 mL
0.8 mL
0.9 mL
1.0 mL
1.1 mL
1.2 mL
1.3 mL
1.4 mL
1.5 mL
1.6 mL
1.7 mL
1.8 mL
1.9 mL
2 mL
2.1 mL
2.2 mL
2.3 mL
2.4 mL
2.5 mL
Diametro (sistema metrico):
6 mm
8 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15889281
Marca: Sampling Systems™ 1500W22
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Developed for customers requiring accurate unit dose samples in small volumes. Sampling Systems™ Tip for Unit Dose Sleeve Sampler is ideal for use with a wide range of free flowing powders and granules.
  • For sampling small volume powders and granules.
  • Chemically resistant 316L stainless steel construction.
  • Different sample sizes can be taken using the same sampler
  • Tips for use with Type 1 or Type 2 Sampling Chambers
Specifica
Product Type Unit Dose Sleeve Sampler Tip
Materiale Stainless Steel (316L)
Capacità (sistema metrico) 2.2 mL
Diametro (sistema metrico) 8 mm
Lunghezza (sistema metrico) 15.5 mm