Learn More
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes
Sfoglia Tutto Epredia Prodotti
Click to view available options
Colore:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
pezzi 1000
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
| Stile pori | Small Squares |
| Chiusura inclusa | Yes |
| Quantità | 1000/Cs. |
| Tipo di chiusura | Separate Lid, Back Hinge |
| Compatibilità con PrintMate | No |
| Colore | White |
| Product Type | TissueLoc Cassette IV with Color-Matched Lids |