Learn More
SI Analytics™ Titration Software
Codice prodotto. 15322437 Sfoglia Tutto SI Analytics Prodotti
Cambia vista
Click to view available options
Product Type: Software
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
15322437 Product Type: Software
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15322437 Fornitore SI Analytics™ N. del fornitore 285220727
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
SI Analytics™ Titration Software is similar to version 3.3 but part 11 compliant.
  • For all TitroLine™ 7XXX titrators and piston burettes Titronic™ 300/500
  • Titration software TitriSoft like Version 3.3, but 21 CFR, part 11 compliant version
  • 3.3P version
TRUSTED_SUSTAINABILITY
Specifica
Tipo Software