Learn More
SI Analytics™ TitroLine™ 7000 Basic Unit Model TL 7000-M1
Sfoglia Tutto SI Analytics Prodotti
Click to view available options
Include:
Magnetic Stirrer TM235
Dimensione della confezione:
Pezzo
Descrizione
- As a rule 10 - 15 user methods are usually enough for the most requirements. But sometimes you need a little bit more capacity. The TitroLine™ 7000 can store up to 50 user methods.
- Highest safety for measuring and calibrating through wireless sensor recognition for TitroLine™ SI Analytics ID-electrodes.
- The new electrodes with definite identification are sending their specific data wireless to the titrator. Therefore the TitroLine™ 7000 uses always the correct calibration data. Erroneous measurements are excluded.
- With a pH stat application a given pH is first adjusted and then kept constant at the certain time with an acid or a base. The pH stat titration is applied to e.g.
- the determination of the enzyme activity
- the pH stat elution of soil sample at pH 4
- the monitoring of the pH value during chemical syntheses
- It is possible to detect and calculate up to 2 equivalence points during one titration. Thanks to this it is possible to determine also the Calcium and Magnesium hardness instead of only the total hardness.
Specifica
Specifica
| Tipo | TitroLine 7000 Basic Unit |
| Include | Magnetic Stirrer TM235 |
| Materiale | Polypropylene |
| Modello | TL 7000-M1 |
| Intervallo temperatura ambiente (sistema metrico) | 10°C to 40°C |
| Lunghezza (sistema metrico) | 45 cm |
| Larghezza (sistema metrico) | 15.3 cm |
| Profondità (sistema metrico) | 29.6 cm |
| Tensione | 100/240 V |