Descrizione
- Can also be used as volumetric KF titrator
- High visible full colour display, that can be easily viewed from a distance and extreme angles
- With intelligent interchangeable modules and storage of all relevant reagent data
- Includes two USB-A, one USB-B, two RS-232 and one LAN port for expansion and connection of devices such as keyboard, printer, barcode reader, memory sticks, balances, PC an further SI Analytics devices
- Storage of results via USB port (PDF and CSV -format) including method transfer
Specifica
Specifica
| Tipi di connettore | Storage of results via USB port (PDF and CSV -format) including method transfer |
| Tipo | Benchtop Titrator |
| Include | Includes two USB-A, one USB-B, two RS-232 and one LAN port for expansion and connection of devices such as keyboard, printer, barcode reader, memory sticks, balances, PC an further SI Analytics devices. |
| Modello | TitroLine 7800 Basic Unit |