Learn More
Invitrogen™ TLR2 Recombinant Rabbit Monoclonal Antibody (8Q6M9)

Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA535274
Descrizione
Immunogen sequence: MPHTLWMVWV LGVIISLSKE ESSNQASLSC DRNGICKGSS GSLNSIPSGL TEAVKSLDLS NNRITYISNS DLQRCVNLQA LVLTSNGINT IEEDSFSSLG SLEHLDLSYN YLSNLSSSWF KPLSSLTFLN LLGNPYKTLG ETSLFSHLTK LQILRVGNMD TFTKIQRKDF AGLTFLEELE IDASDLQSYE PKSLKSIQNV SHLILHMKQH ILLLEIFVDV TSSVECLELR DTDLDTFHFS ELSTGETNSL IKKFTFRNVK ITDESLFQVM KLLNQISGLL ELEFDDCTLN GVGNFRASDN DRVIDPGKVE TLTIRRLHIP RFYLFYDLST LYSLTERVKR ITVENSKVFL VPCLLSQHLK SLEYLDLSEN LMVEEYLKNS ACEDAWPSLQ TLILRQNHLA SLEKTGETLL TLKNLTNIDI SKNSFHSMPE TCQWPEKMKY LNLSSTRIHS VTGCIPKTLE ILDVSNNNLN LFSLNLPQLK ELYISRNKLM TLPDASLLPM LLVLKISRNA ITTFSKEQLD SFHTLKTLEA GGNNFICSCE FLSFTQEQQA LAKVLIDWPA NYLCDSPSHV RGQQVQDVRL SVSECHRTAL VSGMCCALFL LILLTGVLCH RFHGLWYMKM MWAWLQAKRK PRKAPSRNIC YDAFVSYSER DAYWVENLMV QELENFNPPF KLCLHKRDFI PGKWIIDNII DSIEKSHKTV FVLSENFVKS EWCKYELDFS HFRLFDENND AAILILLEPI EKKAIPQRFC KLRKIMNTKT YLEWPMDEAQ REGFWVNLRA AIKS.
TLR2 is a member of the Toll-like receptor (TLR) family which play a fundamental role in pathogen recognition and activation of innate immunity. TLR proteins act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors such as Protein Kinase C (PKC) alpha/beta and NF-kappa-B. TLR2 can form heterodimers with either TLR1 or TLR6, and as a heterodimer, can recognize a variety of bacterial and mycoplasma lipoproteins respectively. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. The various TLRs exhibit different patterns of expression. TLR2 is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB. TLR2 aids in the recognition of pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. Ten human homologs of TLRs (TLR1-10) have been described. TLR2 is expressed in human cells such as tonsils, microglia, lymph nodes, and appendices, activated B-cells in germinal centers. CD14+ monocytes express the highest level of TLR2 followed by CD15+ granulocytes, CD19+ B-cells, and CD3+ T-cells. The expression of TLR2 on different cell types are regulated by different immune response modifiers. For example, LPS, GM-CSF, IL-1, and IL-10 up regulates TLR2 whereas IL-4, IFN-gamma, and TNF down regulate TLR2 expression in monocytes.
Specifica
| TLR2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60603 | |
| Tlr2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-784 of human TLR2 (NP_0032552). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 8Q6M9 | |
| Unconjugated | |
| Tlr2 | |
| CD282; Ly105; TIL4; Tlr2; TLR-2; toll like receptor 2; toll/interleukin-1 receptor-like protein 4; Toll-like receptor; toll-like receptor 2; toll-like receptor 2 variant 1; toll-like receptor 2 variant 2; toll-like-receptor 2, exon 1a_; Toll-like-receptor, exon 1b; Toll-like-receptor, exon 1b_; Toll-like-receptor2, exon 1a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7097 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.