Learn More
TLR7/8 agonist 1 dihydrochloride, MedChemExpress
Codice prodotto. 16480257
Encompass_Preferred
Click to view available options
Quantità:
5 mg
10 mg
25 mg
50 mg
100 mg
Dimensione della confezione:
100mg
10mg
50mg
5mg
Product Code. 16480257
missing translation for 'mfr': Medchem Express HY103698A
MedChemExpress TLR7/8 agonist 1 dihydrochloride is a toll-like receptor TLR7/TLR8 dual-agonistic imidazoquinoline.
TLR7/8 agonist 1 dihydrochloride is a toll-like receptor TLR7/TLR8 dual-agonistic imidazoquinoline.
Specifications
Materiale o nome chimico TLR7/8 agonist 1 dihydrochloride
CAS 1620278-72-9
Colore Light Yellow
Note sul grado di purezza Research
Formula molecolare C22H27Cl2N5
Quantità 50 mg
Informazioni di solubilità DMSO : 83.33 mg/mL (192.72 mM; Need ultrasonic)
SMILES NC1=NC2=CC=CC=C2C3=C1N=C(CCCC)N3CC4=CC=C(CN)C=C4.[H]Cl.[H]Cl
Molecular Weight (g/mol) 432.39
Peso formulazione 432.39
Percent Purity 98.0%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.