Learn More
TLX3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17454-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The TLX3 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TLX3. This antibody recognizes human, mouse, rat antigen. The TLX3 antibody has been validated for the following applications: IP, ELISA analysis.
Specifica
| TLX3 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 30012 | |
| IgG |
| Immunoprecipitation, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |