Learn More
Invitrogen™ TMBIM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5101938
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Antibody detects endogenous levels of total TMBIM1.
Negatively regulates aortic matrix metalloproteinase-9 (MMP9) production and may play a protective role in vascular remodeling.
Specifica
| TMBIM1 | |
| Polyclonal | |
| Unconjugated | |
| TMBIM1 | |
| 2310061B02Rik; AA960455; AU024746; C78899; Lfg3; mKIAA4161; MST100; MSTP100; PP1201; Protein lifeguard 3; Protein RECS1; Protein RECS1 homolog; PSEC0158; RECS1; Responsive to centrifugal force and shear stress gene 1 protein; Tmbib1; Tmbim1; transmembrane BAX inhibitor motif containing 1; transmembrane BAX inhibitor motif-containing protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 316516, 64114, 69660 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BJZ3, Q969X1 | |
| TMBIM1 | |
| A synthesized peptide derived from human TMBIM1(Accession Q969X1), corresponding to amino acid residues M251-T301. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |