missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM70 Mouse anti-Human, Mouse, Clone: 2C11C3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60195-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TMEM70 is a recently identified mitochondrial protein that is thought to play a role in the biogenesis of the ATP synthase in higher eukaryotes. Mutations in this gene result in early neonatal onset of hypotonia, hypertrophic cardiomyopathy, lactic acidos and 3-methylglutaconic aciduria (3-MGC-uria), and usually cause death within the first six weeks of life, although some patients survive much longer. Little is known of the role of TMEM70, but it is conserved across multicellular eukaryotic organisms. It contains a conserved DUF1301 domain and two putative transmembrane regions.Specifica
| TMEM70 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q921N7, Q9BUB7 | |
| TMEM70 | |
| TMEM70 Fusion Protein Ag16947 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2C11C3 | |
| Unconjugated | |
| TMEM70 | |
| TMEM70, transmembrane protein 70 | |
| Mouse | |
| Protein A | |
| RUO | |
| 54968, 70397 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto