Learn More
TNF Mouse, anti-Human, R718, Clone: MAB11, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566958
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
The MAb11 monoclonal antibody specifically binds to human tumor necrosis factor (TNF, also known as TNF-α) protein. TNF is an efficient juxtacrine, paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The MAb11 antibody has been reported to crossreact with Rhesus Macaque TNF.
Specifica
TNF | |
Monoclonal | |
R718 | |
25 Tests | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
MAB11 | |
Mouse | |
RUO | |
Human | |
Liquid |