missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14194-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.Specifica
| TNFSF14 | |
| Polyclonal | |
| Unconjugated | |
| TNFSF14 | |
| CD258, HVEM L, HVEML, LIGHT, LTg, TNFSF14, TR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 301133, 50930, 8740 | |
| -20°C | |
| Liquid |
| ELISA | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43557, Q9QYH9 | |
| Tnfsf14 | |
| TNFSF14 Fusion Protein Ag5380 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto