Learn More
TNIK Mouse, anti-Human, CoraLite 594, Clone: 1E9H3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL594-67948-100UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The TNIK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TNIK. This antibody recognizes human antigen. The TNIK antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™594.
Specifica
| TNIK | |
| 1E9H3 | |
| CoraLite 594 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| KIAA0551, TNIK | |
| 100 μL | |
| 23043 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |