missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TNNC2 Recombinant Protein

Human TNNC2 full-length ORF (AAH05323, 1 a.a. - 160 a.a.) recombinant protein with GST-tag at N-terminal

Marca:  Abnova™ H00007125-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16131472

  • 528.00€ / 25µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Troponin (Tn), a key protein complex in the regulation of striated muscle contraction, is composed of 3 subunits. The Tn-I subunit inhibits actomyosin ATPase, the Tn-T subunit binds tropomyosin and Tn-C, while the Tn-C subunit binds calcium and overcomes the inhibitory action of the troponin complex on actin filaments. The protein encoded by this gene is the Tn-C subunit.

  • Troponin C type 2 (fast)
  • Molecular weight: 43.34kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
  • Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue

Sequence:
MTDQQAEARSYLSEEMIAEFKAAFDMFDADGGGDISVKELGTVMRMLGQTPTKEELDAIIEEVDEDGSGTIDFEEFLVMMVRQMKEDAKGKSEEELAECFRIFDRNADGYIDPEELAEIFRASGEHVTDEEIESLMKDGDKNNDGRIDFDEFLKMMEGVQ

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifica

Specifica

AAH05323
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
43.34
8
Glutathione Sepharose 4 Fast Flow
25 ug
MTDQQAEARSYLSEEMIAEFKAAFDMFDADGGGDISVKELGTVMRMLGQTPTKEELDAIIEEVDEDGSGTIDFEEFLVMMVRQMKEDAKGKSEEELAECFRIFDRNADGYIDPEELAEIFRASGEHVTDEEIESLMKDGDKNNDGRIDFDEFLKMMEGVQ
TNNC2
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
7125
TNNC2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TNNC2
Wheat Germ (in vitro)
GST
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali