missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNNI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22130-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a fast-twitch skeletal muscle protein, a member of the troponin I gene family, and a component of the troponin complex including troponin T, troponin C and troponin I subunits. The troponin complex, along with tropomyosin, is responsible for the calcium-dependent regulation of striated muscle contraction. Mouse studies show that this component is also present in vascular smooth muscle and may play a role in regulation of smooth muscle function. In addition to muscle tissues, this protein is found in corneal epithelium, cartilage where it is an inhibitor of angiogenesis to inhibit tumor growth and metastasis, and mammary gland where it functions as a co-activator of estrogen receptor-related receptor alpha. This protein also suppresses tumor growth in human ovarian carcinoma. Mutations in this gene cause myopathy and distal arthrogryposis type 2B. Alternatively spliced transcript variants have been found for this gene.Specifica
| TNNI2 | |
| Polyclonal | |
| Unconjugated | |
| TNNI2 | |
| AMCD2B, DA2B, FSSV, fsTnI, TNNI2, Troponin I fast skeletal muscle, Troponin I, fast-twitch isoform | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21953, 29389, 7136 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13412, P27768, P48788 | |
| TNNI2 | |
| TNNI2 Fusion Protein Ag17750 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto