missing translation for 'onlineSavingsMsg'
Learn More

GALNT12 Antibody, DyLight 488, R&D Systems™

Product Code. 18048136 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18048136 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18048136 Supplier Novus Biologicals Supplier No. NBP297733G
 Restricted Item
Spedizione stimata: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

GALNT12 Polyclonal specifically detects GALNT12 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT12
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias colorectal cancer, susceptibility to 1, CRCS1, EC 2.4.1.41, FLJ21212, GalNAc-T12, Polypeptide GalNAc transferase 12, polypeptide N-acetylgalactosaminyltransferase 12, pp-GaNTase 12, Protein-UDP acetylgalactosaminyltransferase 12, UDP-GalNAc: polypeptide N-acetylgalactosaminyltransferase, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 12, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 12 (GalNAc-T12)
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GALNT12 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 79695
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.