Learn More
Tocris Bioscience™ GIP (human)
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
Descrizione
GIP (human) is a potent insulinotropic hormone synthesized by duodenal K-cells. High affinity GIP receptor agonist (EC50 = 0.81 nM) that inhibits gastric acid secretion and stimulates pancreatic insulin release in response to glucose. Also affects lipid metabolism and displays mitogenic and antiapoptotic effects in pancreatic β-cells.
Specifica
Specifica
| Purezza | 0.95 |
| Content And Storage | Store at -20°C |
| Quantità | 1 mg |