Learn More
Tocris Bioscience™ TRAP-6
Click to view available options
Quantità:
5 mg
Dimensione della confezione:
5mg
Descrizione
TRAP-6 is a peptide fragment (residues 42-47) of protease-activated receptor 1 (PAR1) that acts as a PAR1 agonist. Stimulates platelet aggregation (EC50 = 0.8 μM), promotes intracellular Ca2+ mobilization and induces rapid phosphodiesterase 3A (PDE3A) phosphorylation in vitro.
Specifica
Specifica
| Purezza | 0.95 |
| Content And Storage | Store at -20°C |
| Quantità | 5 mg |