Learn More
Edmund Bühler GmbH™ Top Frame
Codice prodotto. 10768211
Click to view available options
Da utilizzare con (apparecchiatura):
For SM 2-storey Edmund Bühler
Dimensione della confezione:
Pezzo
Codice prodotto. 10768211
Marca: Edmund Bühler GmbH™ 0052 065
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For use with SM 30 or Combifix shakers. Includes drillings for spring clamps or test tube racks.
  • Top frame for shakers SM 30 with 2 coated trays with drillings for spring clamps or test tube racks
  • Total height: 332mm
  • Distance between the 2 stories: 300mm
  • The rack systems Combifix SM or the universal tray SM can be mounted on the 2-story top frame SM
Specifica
Product Type Top Frame
Da utilizzare con (apparecchiatura) For SM 2-storey Edmund Bühler