Descrizione
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.
Specifica
Specifica
| Product Type | Agarose Gel |