missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53RK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26818-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.Specifica
| TP53RK | |
| Polyclonal | |
| Unconjugated | |
| TP53RK | |
| BUD32, C20orf64, dJ101A2, Nori 2, Nori 2p, p53 related protein kinase, PRPK, TP53 regulating kinase, TP53RK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 112858 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96S44 | |
| TP53RK | |
| TP53RK Fusion Protein Ag25326 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto