missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPR Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA554048
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: KMASVRQHLE ETTQKAESQL LECKASWEER ERMLKDEVSK CVCRCEDLEK QNRLLHDQIE KLSDKVVASV KEGVQGPLNV SLSEEGKSQE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
Steroidogenic factor-1 (SF-1) regulates multiple genes involved in the adrenal and gonadal development and in the biosynthesis of a variety of hormones, including adrenal and gonadal steroids, anti-Mullerian hormone (AMH), and gonadotropins. SF-1 belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors. In the adult ovary, SF-1 localizes to theca/interstitial cells.
Specifica
| TPR | |
| Polyclonal | |
| Unconjugated | |
| TPR | |
| 2610029M07Rik; C77892; Megator; NPC-associated intranuclear protein; nuclear pore complex-associated protein Tpr; nucleoprotein TPR; nucleoprotein TPR-like protein; Tpr; translocated promoter region (to activated MET oncogene); translocated promoter region and nuclear basket protein; Translocated promoter region protein; translocated promoter region, nuclear basket protein; tumor potentiating region | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7175 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12270 | |
| TPR | |
| Recombinant protein corresponding to Human TPR. Recombinant protein control fragment (Product #RP-91828). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto