Learn More
Thermo Scientific™ TRACE™ TR-1 GC Columns
Sfoglia Tutto Thermo Scientific ProdottiDescription
Use this non-polar GC column for method development and analysis of chlorinated and nitroaromatic compounds and environmental samples.
Use the Thermo Scientific™ TRACE™ TR-1 GC column for method development and chlorinated and nitroaromatic compounds and environmental analyses. These non-polar phase, 100% dimethyl polysiloxane gas chromatography columns have a high operating temperature.
Specifications
Specifications
| Temperatura max | 340/360°C |
| Diametro (sistema metrico) interno | 0.25 mm |
| Product Line | TRACE |
| Stationary Phase | TR-1 |
| Tipo USP | G1, G2, G38 |
| Spessore pellicola | 0.25 μm |
| Lunghezza (sistema metrico) | 60 m |
| Polarità | Non polar |