Learn More
Thermo Scientific™ TRACE™ TR-WaxMS GC Columns
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
For high-polarity separations as part of GC/MS analysis, choose the low-bleed, high polarity Thermo ScientificTM TRACETM TR-WaxMS GC Column. The TRACE TR-WaxMS column is manufactured using a proprietary bonding technique that minimizes bleed and extends the maximum operating temperature (260 to 280 ̊C).
- Polar phase, polyethylene glycol
- Proprietary bonding method for expanded operating temperatures
- Extremely low bleed for improved sensitivity and library matches
- High stability with oxygen and water
Applications:
- Aromatic hydrocarbons
- Food additives
- Essential oils
- Alcohols
- Esters
- Aldehydes
- Ketones
Excellent Quality and Reproducibility:
- Low bleed even at elevated temperatures
- Outstanding robustness for difficult separations
- Reliable and reproducible performance
Recommended Uses:
- Aldehydes
- Essential oils
Alternative Uses:
- Acids
- Alcohols
- Alcohols in beverages
- Alditol acetates (sugars)
- Amines&endash;Aliphatic
- Amines&endash;Aromatic
- Herbicides
- Ketones
- Nitroaromatics
- Phenols
- Volatiles
Similar To:
- DB-Wax
- Rtx™-Wax
- Stabilwax™
- HP20M
- BP20
- HP-Wax
- HP-INNOWax
- SUPELCOWAX™ 10
- AT™-Wax
- Nukol™
- CP Wax 52CB
- ZB-Wax
Specifica
Specifica
| Temperatura max | 280/300°C |
| Diametro (sistema metrico) | 0.25 mm |
| Diametro (sistema metrico) interno | 0.25 mm |
| Stationary Phase | Trace™ TG-WAXMS |
| Tipo USP | G16 |
| Da utilizzare con (applicazione) | GC/MS for Aromatic Hydrocarbons, Alcohols, Esters, Aldehydes, and Ketones, GC/MS for Food Additives and Essential Oils |
| Spessore pellicola | 0.5 μm |
| Lunghezza (sistema metrico) | 60 m |
| Polarità | Polar |
| Descrizione | 60m x 0.25mm x 0.5um |