Descrizione
- 100% compatible with all makes and instruments
- Accuracy at 25C is +/- 0.010pH
- Select values manfuactured to IUPAC formulation - features long term buffer stability and buffer resitance to temperature changes during pH meter calibration
- Certified Reference Materials meet Federal/State/local agencies' strictest mandates and deliver exact calibration results for any pH meter.
Specifica
Specifica
| Imballaggio | 2 each 4.005, 7.000, 10.012 100mL bottles |