Learn More
Thermo Scientific™ TraceGOLD™ TG-200MS GC Columns
Codice prodotto. 15810900 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
30 m
60 m
Diametro (sistema metrico) interno:
0.25 mm
0.32 mm
Spessore pellicola:
0.25 μm
0.5 μm
1 μm
Dimensione della confezione:
Pezzo
Codice prodotto. 15810900
Marca: Thermo Scientific™ 260841550
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Resolve compounds that phenyl and cyano phases cannot using a low-bleed GC column with an exceptionally inert phase.
Resolve compounds that phenyl and cyano phases cannot using the Thermo Scientific™ TraceGOLD™ TG-200MS GC Column. The TraceGOLD TG-200MS column has an exceptionally inert phase with selectivity and elution order optimized for difficult separations.
Engineered for Superior Performance
  • Polar phase, trifluoropropylmethyl polysiloxane solid phase to resolve compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including electrochemical detectors, nitrogen phosphorus detectors and mass spectrometers
  • Appropriate for use as a confirmation GC column
Applications:
  • Solvents
  • Fluorocarbons
  • Alcohols and ketones
  • Silanes
  • Glycols
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results, and the low baseline noise enables improved limits of detection with enhanced resolution of low-level analytes.
  • Minimizing column bleed extends the column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended For:
  • Solvents
  • Fluorocarbons
  • Alcohols and ketones
  • Silanes
  • Glycols
  • Alternative Uses:
  • Herbicides
  • Organochlorine pesticides
  • Phenols
  • Similar To:
  • Rtx™-200MS
  • DB-200
  • DB-210
  • TRUSTED_SUSTAINABILITY
    Specifica
    Temperatura max 320/340°C
    Diametro (sistema metrico) 0.32 mm
    Diametro (sistema metrico) interno 0.32 mm
    Product Line TraceGOLD
    Stationary Phase TraceGOLD™ TG-200 MS
    Tipo USP G6
    Da utilizzare con (applicazione) Silanes Analysis, ECD, NPD, and GC/MS for Fluorocarbons, ECD, NPD, and GC/MS for Alcohols and Ketones, ECD, NPD, and GC/MS for Silanes, ECD, NPD, and GC/MS for Glycols
    Spessore pellicola 0.25 μm
    Lunghezza (sistema metrico) 60 m
    Polarità Mid-Polar