Learn More
Thermo Scientific™ TraceGOLD TG-35MS GC Columns
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Thermo Scientific™ TraceGOLD TG-35MS Columns offers a higher phenyl content providing useful elution order and retention time changes.
- Mid-polarity phase, 35% diphenyl/65% dimethyl polysiloxane
- Equivalent to USP G42 phase
- Similar to Rtx™-35, BD-35, HP-35, SPB-35, SPB-608
Recommended for:
- Organochlorine pesticides and herbicides
- Pharmaceuticals
- PCB cogeners
- Aroclor mixes
- Sterols
- Rosin acids
- Phthalate esters
Specifica
Specifica
| Temperatura max | 300/320°C |
| Diametro (sistema metrico) | 0.25 mm |
| Diametro (sistema metrico) interno | 0.25 mm |
| Stationary Phase | TraceGOLD™ TG-35 MS |
| Tipo USP | G42 |
| Da utilizzare con (applicazione) | GC/MS for Organochlorine Pesticides and Herbicides, GC/MS for Pharmaceuticals, GC/MS for PCB Cogeners, GC/MS for Aroclor Mixes, Sterols, Rosin Acids, and Phthalate Esters |
| Spessore pellicola | 1 μm |
| Lunghezza (sistema metrico) | 15 m |
| Polarità | Mid-Polar |
| Descrizione | 1um thickness; 0.25mm ID; 15m length |