Learn More
Thermo Scientific™ TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
The Thermo Scientific™ TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard offer the benefit of an integrated SafeGuard for extended column lifetime.
Thermo Scientific™ TraceGOLD™ TG-5SILMS Guard GC Columns with SafeGuard offer the benefit of an integrated SafeGuard for extended column lifetime.
Using a guard column with a TraceGOLD TG-5SILMS GC column will lead to greater consistency in results over a longer period of time.
- With integrated SafeGUARD
- Available in a range of phases
Specifica
Specifica
| Temperatura max | 330/350°C |
| Diametro (sistema metrico) | 0.25 mm |
| Diametro (sistema metrico) interno | 0.25 mm |
| Product Line | TraceGOLD |
| Stationary Phase | TraceGOLD™ TG-5 SILMS |
| Tipo USP | G27, G36 |
| Da utilizzare con (applicazione) | GC/MS Applications Using Ion-Trap Systems, GC/MS for Polycyclic Aromatics, Phthalates, Phenols, Amines, and Organophosphates, GC/MS for Hydrocarbons Including Chlorinated Hydrocarbons |
| Dimensione particelle | 0.5 μm |
| Gamma di temperatura | -60°C to +350°C |
| Spessore pellicola | 0.5 μm |
| Vedi altri risultati |