Learn More
Thermo Scientific™ TracePLOT™ TG-BOND Alumina GC Columns: KCl Deactivation
Codice prodotto. 10630636 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
30 m
50 m
Diametro (sistema metrico):
0.32 mm
0.53 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10630636
Marca: Thermo Scientific™ 260026110
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use Thermo Scientific™ TracePLOT TG-BOND Alumina GC Columns for linear and quantitative analysis of polar unsaturated hydrocarbons.
Use Thermo Scientific™ TracePLOT TG-BOND Alumina GC Columns for linear and quantitative analysis of polar unsaturated hydrocarbons.
Applications:
  • C1-C5 hydrocarbons
  • Unsaturated hydrocarbon isomers
Engineered for Superior Performance:
  • Strong bonding prevents damage to injection and detection systems from particle release during valve-switching operations.
  • Columns may be regenerated to restore full efficiency and selectivity after water adsorption.
  • Rigorous testing ensures proper film thickness (1,3-butadiene), selectivity (propadiene, methyl acetylene), resolution (trans-2-butene, 1-butene) and coating efficiency (1,3-butadiene).
Similar To:
  • Alumina-PLOT
  • AT™-Alumina
  • CP-Al2O3/KCl
  • CP-Al2O3/Na2SO4
  • GS-Alumina
  • GS-Alumina KCl
  • HP PLOT M
  • HP PLOT S
  • Rt™-Alumina BOND (KCl)
  • Rt™-Alumina BOND (Na2SO4 )
TRUSTED_SUSTAINABILITY
Specifica
Temperatura max Up to 200°C
Diametro (sistema metrico) 0.53 mm
Diametro (sistema metrico) interno 0.53 mm
Stationary Phase TracePLOT™ TG-BOND Alumina
Da utilizzare con (applicazione) Polar Unsaturated Hydrocarbons Analysis, GC for Unsaturated Hydrocarbon Isomers, Linear and Quantitative Analysis of Polar Unsaturated Hydrocarbons
Spessore pellicola 10 μm
Lunghezza (sistema metrico) 50 m