Learn More
Thermo Scientific™ TracePLOT TG-BOND Q+ GC Columns
Codice prodotto. 10249564 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
15 m
30 m
Diametro (sistema metrico):
0.32 mm
0.53 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 10249564
Marca: Thermo Scientific™ 260056030
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Separate ethane, ethylene and acetylene consistently with the intermediate polarity PLOT column with essentially no particle release.
For consistent, reproducible separation of ethane, ethylene, and acetylene, Thermo Scientific™ TracePLOT TG-BOND Q+ GC Columns deliver high performance and unsurpassed stability. The columns use an intermediate-polarity porous divinyl benzene homopolymer phase, and the particles are incorporated into the walls of the tubing for essentially no particle release, which helps prevent damage to detection systems.
Applications:
  • Separation of ethane, ethylene and acetylene to baseline
Similar To:
  • GS-Q
  • Rt™-QS-BOND
TRUSTED_SUSTAINABILITY
Specifica
Temperatura max Up to 250°C
Diametro (sistema metrico) 0.32 mm
Diametro (sistema metrico) interno 0.32 mm
Stationary Phase TracePLOT™ TG-BOND Q+
Da utilizzare con (applicazione) GC for the Separation of Ethane, Ethylene, and Acetylene to Baseline
Spessore pellicola 10 μm
Lunghezza (sistema metrico) 30 m
Polarità Mid-Polar