Learn More
Thermo Scientific™ TracePLOT TG-BOND Q GC Columns
Codice prodotto. 15800900 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
10 m
15 m
30 m
Diametro (sistema metrico) interno:
0.32 mm
0.53 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15800900
Marca: Thermo Scientific™ 260045870
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Achieve consistent, reproducible separation of CO2, methane and 02/N2/CO with the non-polar PLOT column with essentially no particle release.
Achieve consistent, reproducible separation of CO2, methane and 02/N2/CO. Thermo Scientific™ TracePLOT TG-BOND Q GC columns deliver high performance and unsurpassed stability. The columns use a nonpolar, 100% divinyl benzene phase, and the particles are incorporated into the walls of the tubing for essentially no particle release. This strong bonding helps prevent damage to detection systems.
Applications:
  • C1 to C3 isomers and alkanes up to C12
  • Separation of CO2, methane and O2/N2/CO
  • Analysis of oxygenated compounds and solvents
  • Similar To:
  • CP-PoraPLOT Q
  • Rt™-Q-BOND
  • CP-PoraBond Q
  • Supel-Q™-Plot
  • AT™-Q
  • TRUSTED_SUSTAINABILITY
    Specifica
    Temperatura max Up to 300/320°C
    Diametro (sistema metrico) interno 0.32 mm
    Product Line TraceGOLD
    Stationary Phase TracePLOT™ TG-BOND Q
    Da utilizzare con (applicazione) Separation of CO2, Methane, and O2/N2/CO Analysis, Separation of CO2, Methane, and O2/N2/CO, Analysis of Oxygenated Compounds and Solvents
    Spessore pellicola 10 μm
    Lunghezza (sistema metrico) 10 m
    Polarità Non-Polar