Learn More
Thermo Scientific™ TracePLOT TG-BOND U GC Columns
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
15 m
30 m
Diametro (sistema metrico):
0.32 mm
0.53 mm
Dimensione della confezione:
Pezzo
Descrizione
For consistent, reproducible analysis of nonpolar and polar compounds, Thermo Scientific™ TracePLOT TG-BOND U GC Columns deliver high performance and unsurpassed stability. The columns use a polar divinylbenzene ethylene glycol/dimethylacrylate phase, and the particles are incorporated into the tubing for essentially no particle release, which helps prevent damage to detection systems.
Applications:
- Polar and nonpolar compounds
- HP-PLOT U
- Rt™-U-BOND
- CP-PoraPLOT U
- CP-PoraBOND U
- Supel-N PLOT
Specifica
Specifica
| Temperatura max | Up to 190°C |
| Diametro (sistema metrico) | 0.53 mm |
| Diametro (sistema metrico) interno | 0.53 mm |
| Stationary Phase | TracePLOT™ TG-BOND U |
| Da utilizzare con (applicazione) | GC for the Analysis of Polar Compounds |
| Spessore pellicola | 20 μm |
| Lunghezza (sistema metrico) | 15 m |
| Polarità | Polar |