Learn More
TRAK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13770-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.Specifica
| TRAK2 | |
| Polyclonal | |
| Unconjugated | |
| TRAK2 | |
| ALS2CR3, CALS C, GRIF 1, GRIF1, KIAA0549, OIP98, TRAK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171086, 66008, 70827 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60296, Q8R2H7 | |
| Trak2 | |
| TRAK2 Fusion Protein Ag4753 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |