missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TRANCE/TNFSF11/RANK L Antibody (denosumab), Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28639-50ug
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Monoclonal antibody specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TRANCE/TNFSF11/RANK L | |
| Monoclonal | |
| Unconjugated | |
| O14788 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| denosumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| TNFSF11/ RANKL/ CD254 | |
| 50 μg | |
| Cancer | |
| 8600 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction