Learn More
trans-Zeatin-riboside
Codice prodotto. p-7042688
Click to view available options
Quantità:
25 mg
50 mg
100 mg
Dimensione della confezione:
100mg
25mg
50mg
Codice prodotto. 15455629
Marca: Thermo Scientific Alfa Aesar J63382.MB
Richiesta di formato massivo o personalizzato
CAS: 6025-53-2 | C15H21N5O5 | 351.363 g/mol
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
Solubility
Soluble in water, ethanol, and methanol.
Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 6025-53-2
Formula molecolare C15H21N5O5
Molecular Weight (g/mol) 351.363
Numero MDL MFCD00036809
InChI Key GOSWTRUMMSCNCW-HNNGNKQASA-N
Sinonimo zeatin riboside, trans-zeatin riboside, trans-zeatin-riboside, ribosylzeatin, trans-zeatinriboside, 9-ribosyl-trans-zeatin, unii-7lg4d082a9, n-4-hydroxy-3-methyl-2-butenyl adenosine, e-n-4-hydroxy-3-methyl-2-butenyl adenosine, 9-beta-d-ribosyl-trans-zeatin
PubChem CID 6440982
ChEBI CHEBI:71693
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-[[(E)-4-hydroxy-3-methylbut-2-enyl]amino]purin-9-yl]oxolane-3,4-diol
SMILES CC(=CCNC1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)O)O)CO
Specifica
Materiale o nome chimico trans-Zeatin-riboside
Colore White
Quantità 25 mg
Beilstein 5460894
Informazioni di solubilità Soluble in water,ethanol,and methanol.
Peso formulazione 351.36
Forma fisica Powder