missing translation for 'onlineSavingsMsg'
Learn More
Learn More
transgelin/SM22 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2A10C2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60213-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The protein encoded by this gene is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear. Two transcript variants encoding the same protein have been found for this gene.Specifica
| transgelin/SM22 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P31232, P37804, Q01995 | |
| TAGLN | |
| transgelin/SM22 Fusion Protein Ag0764 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse, Pig | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2A10C2 | |
| Unconjugated | |
| TAGLN | |
| SM22, SM22 alpha, SM22a, SM22-a, SM22alpha, Smooth muscle protein 22 alpha, TAGLN, TAGLN1, transgelin, transgelin/SM22 alpha | |
| Mouse | |
| Protein G | |
| RUO | |
| 21345, 25123, 397021, 6876 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto