Learn More
BOLA Transition Fittings
Click to view available options
Lunghezza (sistema metrico):
40 mm
46 mm
Dimensione della confezione:
Pezzo
Descrizione
- Fitting made of PTFE.
- One side with metric thread and connecting nut with compression rings for connecting tubing or tubes with O.D. 4, 6, 8 or 10 mm.
- Other side with female thread UNF ¼ in. 28 G for connecting flanged tubing with O.D. 1.6 or 3.2 mm by means of tube end fittings.
Specifica
Specifica
| Diametro (sistema metrico) esterno | 21 mm |
| Autoclavabile | Autoclavable |
| Lunghezza (sistema metrico) | 40 mm |