Learn More
BOLA Tri-Clamp GL-Adaptor
Click to view available options
Diametro (sistema metrico) interno:
16 mm
32 mm
Dimensione della confezione:
Pezzo
Descrizione
- Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck.
- Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifica
Specifica
| Certificazione/Conformità | FDA |
| Autoclavabile | Autoclavable |
| Diametro (sistema metrico) interno | 32 mm |
| Dimensioni filo | GL 25 |
| Colore | White |