Learn More
Thermo Scientific Chemicals Tris-maleate, 98%
Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Click to view available options
Quantità:
25 g
100 g
Imballaggio:
Bottiglia in vetro
Dimensione della confezione:
100g
25g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Materiale o nome chimico | Tris-maleate |
| Nota nome | 98% |
| Punto di fusione | 104°C to 106°C |
| Colore | White |
| Spettro a infrarossi | Authentic |
| Intervallo percentuale saggio | 98% |
| Imballaggio | Bottiglia in vetro |
| Quantità | 25 g |
| Informazioni di solubilità | Solubility in water: 1g/10 mL |
| Peso formulazione | 237.21 |
| Mehr anzeigen |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur