Learn More
Thermo Scientific™ TSB Blood Bank Refrigerators
Codice prodotto. 17466202 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Tipo di spina:
EU
Capacità (sistema metrico):
140 L
250 L
400 L
700 L
1500 L
Dimensione della confezione:
Pezzo
Codice prodotto. 17466202
Marca: Thermo Scientific™ TSB1500REGBV
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The TSB Series blood bank refrigerators are built with the security, quality, and environmentally sustainable features you have come to expect from Thermo Scientific.
The Thermo Scientific TSB Series Blood Bank Refrigerators are designed to meet the strict requirements for storage of blood and blood related products in full compliance with 93/42/CE – class IIa regulation.
TSB Series Blood Bank Refrigerators are purpose built to meet the stringent guidelines to meet 93/42/CE class IIa regulation as a medical device for storage of blood and blood related products.
  • Wide range of sizes from 140L undercounter to 1500L double door.
  • Touchscreen digital controller with onboard datalogging
  • Whisper quite operation < 50 dBA
  • Triple pane glass doors.
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low energy consumption with real time energy monitoring
  • Low heat output
  • Rear access port
  • Electronic lock with password protection
TRUSTED_SUSTAINABILITY
Specifica
Descrizione TSB Series Blood Bank Refrigerator
Capacità (sistema metrico) 1500 L
Gamma di temperatura 2°C to 8°C
Temperatura (punto di regolazione) +4°C
Refrigerante R290
No. of Doors 2
N. di cassetti 14
Capacità (sistema metrico) ripiano 30 kg
Materiale armadietto Steel
Interno Painted Steel
Certificazione/Conformità 93/42/CEE, 2014/30/UE, 2014/35/UE, 2014/53/UE (only if wireless option), 2011/65/UE
Ruote Yes
Tipo di controllo Touchscreen Control with Password Protection
Porte 1 Rear Access Port
Warranty 36 Months
Sbrinamento Adaptive Defrost
Opzioni di monitoraggio Alarm
Compatibilità Smart View Smart-Vue Pro Compatible
Dimensioni interne (L x P x A) 680 x 1300 x 1500 mm
Dimensioni esterne (L x P x A) 827 x 1440 x 2082 mm
Tipo di spina EU
Requisiti elettrici 230 V/50 Hz
Frequenza 50 Hz
Peso di spedizione (sistema metrico) 283 kg