Learn More
Invitrogen™ TSCOT Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA554147
Codice prodotto. 15752081
  • 560.00€ / 100µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Immunogen sequence: KVPESVAKPS QELPAVDTVS GTVGTYRTLD PDQLDQQYAV GHPPSPGKAK PH Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 62%.
The TSCOT gene, known as the Thymic Stromal Co-transporter, plays a crucial role in the gene expression regulation within thymic epithelial cells, essential for T cell development. It encodes a protein that bears weak homology to bacterial 12 transmembrane co-transporters. Research indicates that TSCOT is predominantly expressed in thymic tissues, although low levels of expression are detectable in other epithelial tissues such as skin and lungs. The gene has been co-analyzed with Foxn1 and Hoxa3, highlighting its significant role in organogenesis. Studies utilizing competitive reverse transcription-polymerase chain reaction (RT-PCR) have mapped TSCOT to specific genomic regions, verifying its expression in a tissue and cell-type specific manner. The introduction of a neoantigen into TSCOT-expressing cells demonstrates its capability to mediate antigen-specific T cell tolerance, offering potential applications in immunological therapies.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
TSCOT
Polyclonal
Unconjugated
SLC46A2
5430429N04Rik; Ly110; putative thymic stromal co-transporter TSCOT; SLC46A2; Solute carrier family 46 member 2; solute carrier family 46, member 2; thymic stromal co-transporter; thymic stromal cotransporter homolog; Thymic stromal cotransporter protein; TSCOT; TSO-1C12
Rabbit
Antigen affinity chromatography
RUO
57864
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BY10
SLC46A2
Recombinant protein corresponding to Human TSCOT. Recombinant protein control fragment (Product #RP-92924).
100 μL