Learn More
TSPAN10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14430-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TSPAN10 belongs to the tetraspanin (TM4SF) family. It is a multi-pass membrane protein. The exact function of TSPAN10 remains unknown.Specifica
| TSPAN10 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN10 | |
| OCSP, Oculospanin, tetraspanin 10, Tspan 10, TSPAN10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83882 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H1Z9 | |
| TSPAN10 | |
| TSPAN10 Fusion Protein Ag5807 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |