Learn More
TPP Techno Plastic Products AG TubeSpin™ Bioreactor
Click to view available options
Capacità (sistema metrico):
15 mL
50 mL
600 mL
Product Type:
Conical, Filter screw cap
Filter Screw Cap
Round, Filter screw cap
Dimensione della confezione:
pezzi 1000
pezzi 180
pezzi 800
Description
- TPP TubeSpin™ Bioreactors have proven their advantages in large scale screening and optimizing of suspension cell system process development.
- The important parameters for the production upscaling can be evaluated with small amounts maximizing the number of parallel experiments at the same time. This lowers the costs and increases the efficiency of the process
- TubeSpin™ Bioreactor 15: working volume 1 - 15 mL, conical or round bottom, filter cap
- TubeSpin™ Bioreactor 50: working volume 1 - 35 mL, conical or round bottom, filter cap
- TubeSpin™ Bioreactor 450: working volume up to approx. 300 ml, conical bottom, filter cap, additional replacement cap available: solid #87355, filter #87356
- TubeSpin™ Bioreactor 600: working volume up to approx. 400 ml, conical bottom, filter cap, additional replacement cap available: solid #87355, filter #87356
- Filter cap with partly 5 different openings (A, B, C, D, E) or 10 equal openings
- Fit standard centrifuge apparatus
- Matching adapter plates for shakers for cultivation of suspension cells available
Specifications
Specifications
| Capacità (sistema metrico) | 600 mL |
| Materiale | Polypropylene |
| Monouso | Single-use |
| Product Type | Conical, Filter screw cap |