Learn More
Scientific Industries SI™ Two-tier Microplate Foam Insert
Click to view available options
Da utilizzare con (apparecchiatura):
Vortex-Genie 2, Vortex-Genie Pulse
Dimensione della confezione:
Pezzo
Descrizione
- High-density, double layered foam insert for prolonged shaking of one standard footprint microplate
- Protects sample from heat-buildup
- Fits into the 152mm Platform attachment (Part No. 15577315)
- For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
Specifica
Specifica
| Capacità | Standard size microplate |
| Materiale | High-density foam |
| Da utilizzare con (apparecchiatura) | Vortex-Genie 2, Vortex-Genie Pulse |
| N. di posizioni | 1 |
| Richiede | Platform attachment 15577315 |