Learn More
Thermo Scientific™ Adattatori provette per rotori TX-200
Codice prodotto. 10799305 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità:
1.5/2.0 mL Conical Microtube
10 mL Blood Collection or 15 mL Corex /Kimble Tube
100 mL Round-bottom Tube
12 x 124 mm Tube
14 mL Conical Urine Tube
15 mL Blood Collection Tube
15 mL Conical Tube
20 mL Round-bottom Tube
25 mL DIN Round-/Flat-bottom Tube
25 mL Universal Tube
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube
50 mL Conical Tube
50 mL DIN Round-bottom Tube
50 mL Round-bottom Tube
Dimensione della confezione:
pezzi 4
Codice prodotto. 10799305
Marca: Thermo Scientific™ 75003809
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm
TRUSTED_SUSTAINABILITY
Specifica
Tipo Rotor Tube Adapter
Capacità 15 mL Blood Collection Tube
Max. Tube Diameter 17 mm
Max. Tube Length 121 mm